Learn More
Cayman Chemical Free Fatty Acid Fluorometric Assay Kit, 96 wells
SDP
Supplier:  Cayman Chemical 70031096WELLS
Encompass_Preferred
The measurement of FFA can be useful in determining metabolic status. Cayman’s Free Fatty Acid (FFA) Assay can be used for measuring free fatty acids in plasma, serum, and urine. The FFA Assay utilizes a coupled enzymatic reaction that results in generation of the highly fluorescent product resorufin.
Catalog No. NC0344245