Learn More
TARGETMOL CHEMICALS INC FFN 206 DIHYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T411955MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. FFN 206 dihydrochloride is a fluorescent VMAT2 substrate that can be used to detect VMAT2 subcellular sites in cell culture and shows no detectable inhibition of DAT. purity: 98%
Catalog No. 50-257-9951