Learn More
Biochain Institute Inc FFPE and Frozen Matched Pair Genomic DNA: Human Tumor Tissue: Lung, 2 x 2 ug/pk
SDP
Supplier:  Biochain Institute Inc D8235152FP
Encompass_Preferred
Matched Pair products include: Primary Pair (PP), or Primary and metastatic Pair (PM). PP consists of a pair of genomic DNA isolated from primary tumor and its adjacent normal tissue; PM consists of a pair of genomic DNA from primary tumor and corresponding metastatic tumor. Genomic DNA in each pair is prepared from the same donor. This product line is designed for identifying tumor-specific genes and tumor metastatic.
Catalog No. 50-187-1526