Learn More
Selleck Chemical LLC FG-2216
SDP
Supplier:  Selleck Chemical LLC S79795MG
Encompass_Preferred
FG-2216 is a potent and orally active HIF prolyl 4-hydroxylase inhibitor with IC50 of 3 9 M for PHD2 Phase 2
Catalog No. 50-000-01181