Learn More
Thermo Scientific Beta Cyanoethyl Phosphoramidite, dC(Bz), 102212-98-6, MFCD00036315, 2 g
Supplier:  Thermo Scientific 401160
Encompass
MF: C46H52N5O8P, MW: 833.918, Chemical Name: dC(Bz) beta CEP. The dC(Bz) Beta Cyanoethyl Phosphoramidite is used as a component of a deprotecting chemistry set including dA(Bz), dG(iBu) and T. Standard series of phosphoramidite deprotects in 6-8 hours at 55°C.
Catalog No. NC0382655
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.