Learn More
Thermo Scientific CE Phosphoramidite, dC(Bz), 0.5 g
Supplier:  Thermo Scientific 400332
Encompass
Chemical Name: dC(Bz) CEP. The dC(Bz) CE Phosphoramidite is used for nucleic acid synthesis on ABI DNA Synthesizers. Standard series of phosphoramidite deprotects in 6-8 hours at 55°C.
Catalog No. NC0382652
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov