Learn More
Thermo Scientific CE Phosphoramidite, dG(iBu), 93183-15-4, MFCD00055147, 0.5 g
Supplier:  Thermo Scientific 400331
Encompass
MF: C44 H54 N7 O8 P, MW: 839.926, Chemical Name: dG(iBu) CEP. The dG(iBu) CE Phosphoramidite is used for nucleic acid synthesis on ABI DNA Synthesizers. Standard series of phosphoramidite deprotects in 6-8 hours at 55°C.
Catalog No. NC0382651
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov