Learn More
Thermo Scientific FG G DMF AMIDITE 4.59G
Supplier:  Thermo Scientific 4339777
Encompass
5'-O-Dimethoxytrityl-N2(dimethylaminoformamidine)-2'-deoxyguanosine-3'-O-( beta -cyanoethyl-N,N-diisopropyl) phosphoramidite
Catalog No. NC0382663
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov