Learn More
Medchemexpress LLC FGF-1 Mouse 1mg

Supplier: Medchemexpress LLC HYP70651MG
Recombinant mouse fibroblast growth factor-1 supplied as a lyophilized, tag-free protein for in vitro studies of cell proliferation, angiogenesis, and signaling. The preparation is validated for purity and bioactivity and is intended for use in cell-based assays requiring biologically active FGF-1.
- High purity (>95%) as determined by reducing SDS-PAGE.
- Lyophilized powder for long-term stability and convenient storage.
- Tag free to minimize interference in binding or signaling assays.
- Predicted molecular mass ~15.8 kDa; observed ~18 kDa by SDS-PAGE.
- High specific activity (ED50 ≈ 1-2 ng/mL in fibroblast proliferation assays).
- Stable at -20°C; reconstituted material stable short term at 4°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.