Learn More
Medchemexpress LLC Fibroblast growth factor 21 | 500UG

Supplier: Medchemexpress LLC HYP701092500UG
Recombinant human fibroblast growth factor 21 (FGF-21), 181 amino acids, supplied as a lyophilized powder. FGF-21 is an endocrine regulator of energy homeostasis with reported metabolic and cardioprotective effects. This product sheet provides formulation, endotoxin, activity, and storage information for research applications.
- Sequence length: 181 amino acids.
- Molecular weight: ~23 kDa (SDS-PAGE).
- Endotoxin: <1 EU/μg (LAL).
- Formulation: lyophilized from 20 mM PB, 150 mM NaCl or PBS with 8% trehalose.
- Expression system: Escherichia coli.
- Biological activity: ED50 ≤1.071 μg/mL (NIH-3T3 assay with β-Klotho).
- Storage: store at -20°C; reconstituted stable at 4°C for 1 week or -20°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.