Learn More
Enzo Life Sciences FGF-8 (human), (recombinant) (25µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1740025
Encompass_Preferred
Alternative name: Fibroblast growth factor-8, FGF-8b, AIGF, HBGF-8. Purity: ≥95% (SDS & HPLC). Formulation: Lyophilized from 10mM sodium phosphate, pH 8.0, containing 75mM NaCl. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-3963