Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGF basic/bFGF Huma 250ug
SDP

Supplier:  Medchemexpress LLC HYP70600A250UG

Encompass_Preferred

Recombinant human basic fibroblast growth factor (bFGF) K128N variant supplied lyophilized and tag-free for research applications. The protein is expressed in E. coli, purified by reducing SDS-PAGE to high purity, and provided with recommended reconstitution and storage instructions to support consistent experimental use.

  • Lyophilized, tag-free recombinant protein for research applications.
  • E. coli expression ensures consistent recombinant production.
  • Purity greater than 95% as assessed by reducing SDS-PAGE.
  • Apparent molecular weight ~18 kDa (predicted 17.2 kDa).
  • Reconstitution recommended at ≥100 μg/mL to achieve working concentrations.
  • Stable long-term at -20°C; aliquot and store at -20°C or -80°C for extended storage.
  • Formulated with trehalose and mannitol to improve stability during lyophilization.

Catalog No. 50-001-84606


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.