Learn More
ABclonal Technology FGF2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2233020UL
Encompass_Preferred
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family FGF family members bind heparin and possess broad mitogenic and angiogenic activities This protein has been implicated in diverse biological processes such as limb and nervous system development wound healing and tumor growth The mRNA for this gene contains multiple polyadenylation sites and is alternatively translated from non-AUG (CUG) and AUG initiation codons resulting in five different isoforms with distinct properties The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect whereas the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF
Catalog No. 50-000-06492