Learn More
Medchemexpress LLC FGFR-4 Human HEK29 50ug
SDP
Supplier:  Medchemexpress LLC HYP7305950UG
Encompass_Preferred
The FGFR-4 protein is a tyrosine protein kinase and cell surface receptor for fibroblast growth factor that regulates diverse cellular processes such as proliferation differentiation migration lipid metabolism and bile acid biosynthesis Notably it contributes to the downregulation of the bile acid synthase CYP7A1 in response to FGF19 FGFR-4 Protein Human (HEK293 His) is the recombinant human-derived FGFR-4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80501