Learn More
Medchemexpress LLC FGFR-4 Rat HEK293 100ug
SDP
Supplier:  Medchemexpress LLC HYP73061A100UG
Encompass_Preferred
The FGFR-4 protein is a tyrosine-protein kinase receptor for fibroblast growth factor that regulates cellular processes lipid metabolism bile acid biosynthesis and more Its effects include downregulation of CYP7A1 in response to FGF19 FGFR-4 Protein Rat (HEK293 C-hFc) is the recombinant rat-derived FGFR-4 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80502