Learn More
Epigentek Inc FGFR3 Polyclonal Antibody, Biotin Conjulated (50 ul)
SDP
Supplier:  Epigentek Inc A53800050
Encompass_Preferred
Biotin Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Fibroblast growth factor receptor 3 protein (23-375AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: CD_antigen=CD333 FGFR3 JTK4.
Catalog No. 50-122-1783