Learn More
Medchemexpress LLC Fgi-106 tetrahydrochloride | 1149348-10-6 | 99.7% | 604.49 g/mol | C28H42Cl4N6 | 50MG
SDP
Supplier:  Medchemexpress LLC HY124618A50MG
Encompass_Preferred
FGI-106 tetrahydrochloride is a research-grade, broad-spectrum antiviral small molecule (CAS 1149348-10-6) supplied as the tetrahydrochloride salt. It is reported to inhibit multiple viruses including Ebola, Rift Valley fever virus, Dengue virus, HCV, and HIV-1, and is intended for in vitro antiviral and mechanistic studies.
  • Broad-spectrum antiviral activity against multiple viral families.
  • Supplied as the tetrahydrochloride salt for improved handling and solubility.
  • High purity suitable for research use (~99.7%).
  • Molecular formula C28H42Cl4N6; molecular weight 604.49 g/mol.
  • Available in multiple milligram-scale package sizes for assay flexibility.
Catalog No. 50-002-37145