Learn More
ABclonal Technology FGL2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1375520UL
Encompass_Preferred
The protein encoded by this gene is a secreted protein that is similar to the beta- and gamma-chains of fibrinogen. The carboxyl-terminus of the encoded protein consists of the fibrinogen-related domains (FRED). The encoded protein forms a tetrameric complex which is stabilized by interchain disulfide bonds. This protein may play a role in physiologic functions at mucosal sites.
Catalog No. 50-001-14961