Learn More
Apexbio Technology LLC FH535, 10mg. Cas: 108409-83-2 MFCD: MFCD01212888. Wnt/B-catenin inhibitor
SDP
Supplier:  Apexbio Technology LLC A341310
Encompass_Preferred
FH535 is a small molecule inhibitor of Wnt/B-catenin with IC50 values of 15.4M, 10.9M, 9.3M, respectively in LCSC, Huh7, PLC cell lines [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4800