Learn More
TARGETMOL CHEMICALS INC FH535 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T241350MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FH535 a Wnt/(beta)-catenin signaling and PPAR inhibitor exhibits anti-tumor activities. purity: 99%
Catalog No. 50-258-0830