Learn More
Medchemexpress LLC HY-144835 5mg , FHD-286 CAS:2671128-05-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1448355MG
Encompass_Preferred
HY-144835 5mg FHD-286 CAS: 2671128-05-3 FHD-286 is a BRG1/BRM ATPase inhibitor for the treatment of BAF-related disorders such as acute myeloid leukemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0129