Learn More
Medchemexpress LLC FHT 2344 | 2468058-90-2 | >95.0% | 528.6 g·mol⁻1 | C23H24N6O5S2 | 10 MG

Supplier: Medchemexpress LLC HY14945810MG
FHT-2344 is a small-molecule chemical probe that potently inhibits the ATPase activity of SMARCA4 (BRG1) and SMARCA2 (BRM). It has been characterized in preclinical studies with anticancer activity and is supplied as a high-purity solid or as solutions for in vitro use.
- Potent dual SMARCA2/SMARCA4 ATPase inhibition (SMARCA2 IC50 ≈ 0.013 μM; SMARCA4 IC50 ≈ 0.026 μM).
- High selectivity versus CHD4 (no significant inhibition at ≤200 μM).
- Validated in cell and xenograft models for transcriptional and antitumor effects.
- Available as solid powder or 10 mM solution in DMSO for convenience.
- High purity ≥95% by HPLC.
- Molecular formula C23H24N6O5S2; molecular weight 528.6 g·mol⁻1.
- Store at -20°C to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.