Learn More
Electron Microscopy Sciences Double FIB Sample and Grid Holder, Pin, 1/2"
SDP
Catalog No. 501927129
Encompass_Preferred
Holds a FIB sample mounted on two standard 1/2" (12.7 mm) pin stubs for FIB milling and lift-out procedures. Conveniently holds two FIB grids close to the sample to mount prepared TEM lamellae on the FIB grid for TEM imaging. Cost-effective holder suitable for all FIB/SEM systems which accept pin mount holders, including the FEI, ZEISS and Tescan systems. For the JEOL and Hitachi systems, use a pin mount adapter.
Overall Dimensions: 36.5 x 12.7 x 11.6 mm (1.44" x 0.5" x 0.46").
Pin: Standard 3.2 mm ( 1/8").
Material: Vacuum grade aluminum with brass screws.
Includes Philips screwdriver #0.
Catalog No. 50-192-7129 Supplier Electron Microscopy Sciences Supplier No. 7595004
Holds a FIB sample mounted on two standard 1/2" (12.7 mm) pin stubs for FIB milling and lift-out procedures. Conveniently holds two FIB grids close to the sample to mount prepared TEM lamellae on the FIB grid for TEM imaging. Cost-effective holder suitable for all FIB/SEM systems which accept pin mount holders, including the FEI, ZEISS and Tescan systems. For the JEOL and Hitachi systems, use a pin mount adapter.
Overall Dimensions: 36.5 x 12.7 x 11.6 mm (1.44" x 0.5" x 0.46").
Pin: Standard 3.2 mm ( 1/8").
Material: Vacuum grade aluminum with brass screws.
Includes Philips screwdriver #0.