Learn More
Grainger Fiber Cord Duplex ST ST 1m Orange
Supplier:  Grainger 14C123
Encompass_Preferred
The Grainger 14C123 is a Westward brand 1/4-inch drive click-type torque wrench with a range of 10 to 150 inch pounds. It has a reversible ratchet head and comes with a molded plastic case. The torque wrench has an accuracy of ±4% clockwise and has a chrome vanadium steel construction with a satin chrome finish. It measures 15-1/2 inches long and weighs 1.95 pounds. This torque wrench is ideal for precise tightening applications.
Catalog No. 50-275-2960