Learn More
Cole-Parmer Autoclaveable White Fiberglass Tray, 12 x 13 x 3/4"
Supplier:  Cole-Parmer 0671562
Encompass_Preferred
Autoclaveable White Fiberglass Tray, 12 x 13 x 3/4", Rugged fiberglass trays withstand temperatures 0 to 180°F (-17 to 83°C) under normal usa. Autoclavable to 500°F (204°C) for short intervals.
Catalog No. NC1091401