Learn More
Medchemexpress LLC Fibrinogen-Binding Peptide TFA | 99.32% | 679.64 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP1741A5MG
Encompass_Preferred
Fibrinogen-Binding Peptide TFA (designed by anticomplementarity hypothesis) is a presumptive peptide mimic of the vitronectin binding site on the fibrinogen receptor. Fibrinogen-Binding Peptide TFA binds fibrinogen and inhibits both the adhesion of platelets to fibrinogen and platelet aggregation, and also inhibits the adhesion of platelets to vitronectin.
  • Binds fibrinogen
  • Inhibits adhesion of platelets to fibrinogen
  • Inhibits platelet aggregation
  • Inhibits adhesion of platelets to vitronectin
  • Presumptive peptide mimic of the vitronectin binding site on the fibrinogen receptor
Catalog No. 50-004-22629