Learn More
Enzo Life Sciences Fibrinogen (human plasma) (100mg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1570100
Encompass_Preferred
Alternative name: Factor 1. Purity: ≥95%. Formulation: Liquid. In 20mM sodium citrate-HCl, pH 7.4. Source: Isolated from human plasma. Plasminogen depleted. Long Term Storage: -80°C.
Catalog No. 50-201-3946