Learn More
Sigma Aldrich Fine Chemicals Biosciences Fibrinogen from bovine plasma, 9001-32-5, MFCD00163624, 5 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F86305G
Encompass
Type I-S, 65-85% protein (≥75% of protein is clottable), storage temp: −20°C, Synonym: Factor I. Fibrinogen has been used in studies of haemostatic therapy in surgical and massive trauma patients. These studies have shown that fibrinogen may prove to be more superior in stopping blood loss when compared to using fresh frozen plasma (FFP).
Catalog No. NC1042134