Learn More
CPC Scientific Glp-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific FIBN003B
Encompass
Peptide released from fibrinogen on thrombin action. This action appears to be necessary for Ca2+ uptake and its subsequent binding to fibrin.ONE-LETTER SEQUENCE: Glp-GVNDNEEGFFSARMOLECULAR FORMULA: C66H93N19O25MOLECULAR WEIGHT:1552.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [36024-23-6], RESEARCH AREA: Hematology
Catalog No. 50-210-9276