Learn More
Medchemexpress LLC Fibrinopeptide B, human TFA | 99.2% | 1666.58 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP1493A5MG
Encompass_Preferred
Fibrinopeptide B, human TFA (FPB,human TFA) is a 14-amino acid peptide, which is released from the amino-terminus of β-chains of fibrinogen by thrombin. It is intended for research use only.
  • Released from amino-terminus of β-chains of fibrinogen by thrombin
  • Causes directed cell migration of neutrophils (PMN) and fibroblasts
  • Possesses potent chemotactic activity relative to human complement (C5a), leukotriene B4 (LTB4), and formyl-methionyl-leucyl-phenylalanine (fMLP)
  • Chemotactic activity for fibroblasts is comparable to that of platelet-derived growth factor
  • Not chemotactic for monocytes
  • Shows no interaction with PMN receptor for C5a, LTB4, or fMLP
Catalog No. 50-004-24313