Learn More
Sigma Aldrich Fine Chemicals Biosciences Fibronectin human plasma, 86088-83-7, MFCD00131062, 1 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F20061MG
Encompass
Lyophilized powder, BioReagent, suitable for cell culture, Synonym: Fibronectin. Fibronectin from human plasma is used for the following applications: Used in cell seeding and neuronal differentiation (as one of the components in expansion medium for the re-suspension of CBSCs (Cord blood stem cells) and is also used in differentiation medium). Used in cell culture (the culture chamber was coated with fibronectin for the expression of HEK293 cells). Used in murine embroyonic stem cell line (CCE) culture and induction of endothelial cell differentiation (for the induction of endothelial cell differentiation, cultured cells were pre-coated with fibronectin).
Catalog No. NC0168080