Learn More
Promocell GmbH FIBRONECTIN SOL HUMAN 5ML
Supplier:  Promocell GmbH C43060
Encompass
Cell Media Supplements, PromoCell, Fibronectin Solution, human (1 mg/ml), cell culture tested, 5ML. PromoCell Fibronectin is a clear sterile solution obtained by affinity purification on gelatin-sepharose from irradiated citrated bovine or human plasma, respectively.
Catalog No. 50-306-410