Learn More
Selleck Chemical LLC Fidaxomicin 50mg 873857-62-6 OPT-80, PAR-101
SDP
Supplier:  Selleck Chemical LLC S422750MG
Encompass_Preferred
Fidaxomicin (OPT-80, PAR-101) is a narrow spectrum macrocyclic antibiotic that inhibits RNA polymerase sigma subunit. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3785