Learn More
Medchemexpress LLC Fidaxomicin | 873857-62-6 | 99.8% | 1058.04 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17580R50MG
Encompass_Preferred
Fidaxomicin is a macrocyclic antibiotic that acts as an orally active and potent RNA polymerase inhibitor. It exhibits a narrow spectrum of antibacterial activity and is particularly effective against *Clostridium difficile* (MIC90=0.12 μg/mL), making it suitable for research related to *Clostridium difficile* infection (CDI).
  • Selectively eradicates pathogenic *Clostridium difficile* with minimal disruption to normal, healthy intestinal flora.
  • Minimally impacts commensals and does not inhibit commonly cultured bowel commensals (MIC90 >1024 μg/mL).
  • Completely prevents lethality and relapses in a hamster model for pseudomembranous colitis.
  • Prevents development of antibiotic-induced *C. difficile* colitis in hamsters at doses as low as 0.2 mg/kg.
Catalog No. 50-003-87344