Learn More
Medchemexpress LLC Fidrisertib | 2141955-96-4 | 98.2% | 562.70 g/mol | C31H42N6O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11788710MG
Encompass_Preferred
Fidrisertib is an orally active small-molecule inhibitor of activin receptor-like kinase 2 (ALK2) used as a research reagent to study ALK/TGF-β signaling and related disease models. The compound is provided as a solid research chemical with indicated purity suitable for in vitro and preclinical research applications.
  • Orally active ALK2 inhibitor, useful for studies of ALK/TGF-β pathways.
  • Supplied as a solid research reagent with high reported purity.
  • Relevant for oncology and fibrodysplasia ossificans progressiva research models.
  • Soluble in common organic solvents such as DMSO for assay preparation.
  • Provided in small research pack suitable for assay development and preclinical experiments.
Catalog No. 50-002-28137