Learn More
Cole-Parmer Rite in the Rain 550F Bound Environmental Notebook; 1/Pk
Supplier:  Cole-Parmer 0657000
Encompass_Preferred
The Rite in the Rain bound notebooks are crafted with sturdy polydura covers. The notebooks measure 4-34 by 7-12 inches and consist of 156 pages or 78 sheets that are numbered. The environmental book meets US EPA guidelines. They contain 10 helpful reference pages with a field page pattern.
Catalog No. NC9729667