Learn More
TARGETMOL CHEMICALS INC FIIN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T374265MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. FIIN-1 (FGFR irreversible inhibitor-1) is an irreversible and selective FGFR inhibitor with Kd of 2.8 6.9 5.4 120 32 and 120 nM for FGFR1 FGFR2 FGFR3 FGFR4 Flt1 Flt14 respectively.FIIN-1 inhibited FGFR1 FGFR2 FGFR3 FGFR4 with IC50 of 9.2 6.2 11.9 and 189 nM. FIIN-1 inhibited FGFR1 FGFR2 FGFR3 and FGFR4 with IC50s of 9.2 6.2 11.9 and 189 nM respectively. purity: 97%
Catalog No. 50-258-0139