Learn More
Selleck Chemical LLC FIIN-2
SDP
Supplier:  Selleck Chemical LLC S77145MG
Encompass_Preferred
FIIN-2 is an irreversible pan-FGFR inhibitor with IC50 of 3 09 nM 4 3 nM 27 nM and 45 3 nM for FGFR1/2/3/4 respectively
Catalog No. 50-217-3298