Learn More
Cayman Chemical FIlIpIn II 2.5mg
SDP
Supplier:  Cayman Chemical 255212.5 MG
Encompass_Preferred
An active component of the filipin complex; reduces growth of various fungi (MICs = 1-10 μg/ml); inhibits ADP-stimulated respiration in rat liver mitochondria at 3.5 mg/ml; induces hemolysis of liposomes in a cholesterol-independent manner (IC50 = 0.5 μg/ml)
Catalog No. 50-301-5881