Learn More
Apexbio Technology LLC Filipin III, 1mg. Cas: 480-49-9 MFCD: MFCD30742956. cholesterol-binding, fluorescing antibiotic used for the detection of lipoproteins
SDP
Supplier:  Apexbio Technology LLC B60341
Encompass_Preferred
Filipin is the collective name given to four isomeric polyene macrolides isolated from cultures of S. filipinensis; Filipin III is the predominant isomer and the one used in most studies. Filipin binds to cholesterol in membranes, forming ultrastructural aggregates and complexes which can be visualized by freeze-fracture electron microscopy [1,2]. The binding of cholesterol also decreases the intrinsic fluorescence of Filipin, and this property has also been used to detect cholesterol in membrane fractions. Other sizes are available. Please inqury us for quote.
Catalog No. 50-115-2556