Learn More
Research Products International Corp Film Sealing Paddle, 1 each
SDP
Supplier:  Research Products International Corp 248743
Encompass
Film-Sealing Paddles are an ideal alternative to using a plate roller. Paddle facilitates pressing films to ensure a secure uniform seal around all wells on 96-well, 384-well, and 1536-well plates. Paddles are also recommended for sealing films on raised-rim plates.
Catalog No. 50-223-0148