Learn More
Grainger Film Tape 1/2 in x 5 yd Amber 2.5 mil
Supplier:  Grainger 15C590
Encompass_Preferred
The TAPECASE Film Tape 1/2 in x 5 yd is a general purpose masking tape made of crepe paper with a rubber-based adhesive. It has a temperature resistance up to 200°F and can be used for holding, sealing, splicing, masking and wrapping. This tape tears easily by hand without curling and conforms well to irregular surfaces. It is offered in multiple widths and lengths. This particular tape is 1/2 inch wide and 5 yards long. The rubber adhesive provides instant adhesion that resists lifting or curling and removes cleanly after use.
Catalog No. 50-275-1798