Learn More
Grainger Film Tape 1 in x 1 in Amber 2.5 mil PK28
Supplier:  Grainger 16U723
Encompass_Preferred
The TapeCase Film Tape is a brown, 2-inch wide, pressure-sensitive packaging tape made in the USA. It features a natural rubber adhesive and a 3.6 mil biaxially oriented polypropylene film backing. This tape is designed for box sealing, splicing, and general packaging applications. It adheres instantly to corrugated fiberboard and Kraft surfaces. The tape has a tensile strength of 25 lbs/in width and can withstand temperatures between -20°F to 160°F. It meets the ASTM D5330 standard for packaging tapes. The TapeCase Film Tape comes in a convenient hand-held dispenser with serrated edge for easy tear.
Catalog No. 50-276-7516