Learn More
Grainger Film Tape 2 in x 2 in Amber 2.5 mil PK15
Supplier:  Grainger 16U724
Encompass_Preferred
The TapeCase Film Tape is a brown, 2-inch wide, pressure-sensitive tape made of polypropylene film backing and rubber adhesive. It is designed for packaging, sealing, splicing, and general purpose use. This tape has moderate adhesion and is resistant to UV rays, moisture, and chemicals. It can be used indoors or outdoors and works well at temperatures ranging from -20°F to 200°F. The tape is available in various lengths, with a standard length of 60 yards per roll.
Catalog No. 50-276-6468