Learn More
Grainger Film Tape 2 in x 5 yd Amber 2.5 mil
Supplier:  Grainger 15C593
Encompass_Preferred
The Grainger 15C593 film tape is a 2 inch wide polyimide amber tape that comes in 5 yard rolls. It has a maximum operating temperature of 500°F and is made in the USA. This high temperature resistant tape provides electrical insulation and can be used for harness wrapping, banding, and identification purposes. With its conformability and thermal stability, this tape is well-suited for applications in the aerospace, automotive, and electronics industries.
Catalog No. 50-274-9544