Learn More
Grainger Film Tape 3/4 in x 5 yd Amber 2.5 mil
Supplier:  Grainger 15C591
Encompass_Preferred
The TAPECASE Film Tape 3/4 in x 5 yd is a general purpose masking tape made of crepe paper with a rubber-based adhesive. It has a tensile strength of 18 lbs/in width and adhesion to steel of 25 oz/in width. The tape is 0.75 inches wide and 5 yards long per roll. It can be used for holding, sealing, splicing, masking and other general purposes. The tape has good conformability and adhesion to irregular surfaces.
Catalog No. 50-274-9653