Learn More
Grainger Film Tape 3/4inx3/4in Amber 2.5mil PK42
Supplier:  Grainger 16U722
Encompass_Preferred
The Grainger Approved Film Tape is a 2" wide clear polypropylene film tape with an acrylic adhesive. It is designed for general purpose holding, splicing, sealing and masking applications. The tape has a temperature range of up to 200°F and offers quick stick at application. It can be used for packaging, shipping, industrial assembly, arts and crafts, and more. The tape is UL listed and comes in a convenient 60 yard roll for efficient use.
Catalog No. 50-276-6656