Learn More
Grainger Film Tape 4 in x 36 yd Amber 2.5 mil
Supplier:  Grainger 15C602
Encompass_Preferred
The TAPECASE Film Tape 4 in x 36 yd is a 4 inch wide by 36 yard long clear polypropylene film tape. It features a pressure sensitive acrylic adhesive and has a tensile strength of 22 lbs/in. The tape is designed for general purpose packaging, sealing, holding, mending, splicing, and masking applications. It can adhere to paper, plastic, glass, wood, and metal surfaces. The operating temperature range is 32 to 150 degrees Fahrenheit.
Catalog No. 50-275-0643