Learn More
Premier Lab Supply FilmVelopes, PN# FV-160-212 – Mylar film 6.0μm (0.24mil)/2.0” Diameter
SDP
Supplier:  Premier Lab Supply FV160212
Encompass
FilmVelopes - CLEAN, EASY & FASTER sample prep without the tear away! (PN# FV-160-212). Innovative proprietary thin film applicator utilizing specialty jackets firmly holding a thin film in place while allowing for a clean release upon application. FilmVelopes reduces the probability of error and excess film waste while allowing for easy handling. 100 per pack.
Catalog No. NC1193343