Learn More
Chemglass Life Sciences Complete Filter Apparatus, Coarse Porosity
SDP
Supplier:  Chemglass Life Sciences CG140403
Encompass_Preferred
Complete Filter Apparatus, Coarse Porosity. Useful for direct filtration applications of small volumes under vacuum. Complete apparatus consists of a Buchner funnel with 14/20 top outer and lower inner joints, and a 50mL Erlenmeyer flask having a 14/20 outer joint.
Catalog No. CG140403